Learn More
Troxerutin (85%), TRC
Product Code. 30456975
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456975 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456975 Supplier Toronto Research Chemicals Supplier No. TRCT89320050MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Troxerutin
CAS 7085-55-4
Molecular Formula C33 H42 O19
Quantity 50 mg
Synonym 2-[3,4-Bis(2-hydroxyethoxy)phenyl]-3-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-7-(2-hydroxyethoxy)-4H-1-benzopyran-4-one, 3,5-dihydroxy-3',4',7-tris(2-hydroxyethoxy)flavone 3-[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside], 3,5-Dihydroxy-3',4',7-tris(2-hydroxyethoxy)flavone 3-rutinoside, 3',4',7-Tri-O-(β-hydroxyethyl)rutoside, 3',4',7-Tris(hydroxyethyl)rutin, Factor P-Zyma, Posorutin, Rufen-P4, Ruven, THR, TriHER, Trioxyethylrutin, Tris(hydroxyethyl)rutin, Tris(hydroxyethyl)rutoside, Tris-O-(2-hydroxyethyl)rutin, Tris-O-(β-hydroxyethyl)rutoside, Troxerutin, Troxerutine, Troxevazin, Vastribil, Veinamitol, Veniten, Venoruton P4, Vitamin P4, Z 6000
InChI Formula InChI=1S/C33H42O19/c1-14-23(38)26(41)28(43)32(49-14)48-13-21-24(39)27(42)29(44)33(51-21)52-31-25(40)22-17(37)11-16(45-7-4-34)12-20(22)50-30(31)15-2-3-18(46-8-5-35)19(10-15)47-9-6-36/h2-3,10-12,14,21,23-24,26-29,32-39,41-44H,4-9,13H2,1H3/t14-,21+,23-,24+,26+,27-,28+,29+,32+,33-/m0/s1
SMILES C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(OCCO)cc(O)c4C3=O)c5ccc(OCCO)c(OCCO)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
IUPAC Name 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
Molecular Weight (g/mol) 742.68
Formula Weight 742.232