Learn More
Thermo Scientific™ TSB Blood Bank Refrigerators
Codice prodotto. 17486212 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Tipo di spina:
EU
Capacità (sistema metrico):
140 L
250 L
400 L
700 L
1500 L
Dimensione della confezione:
Pezzo
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo di spina Capacità (sistema metrico) Dimensione della confezione
17486212 EU 700 L Pezzo
17426202 EU 140 L Pezzo
17466202 EU 1500 L Pezzo
17406212 EU 250 L Pezzo
17446212 EU 400 L Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Codice prodotto. 17486212 Fornitore Thermo Scientific™ N. del fornitore TSB700REGBV
  • ONLINE EXCLUSIVE PRICE
    10241.96€
    9400.00€ / pezzi 1
    Risparmia 841.96€
    8% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 11-03-2027
The TSB Series blood bank refrigerators are built with the security, quality, and environmentally sustainable features you have come to expect from Thermo Scientific.
The Thermo Scientific TSB Series Blood Bank Refrigerators are designed to meet the strict requirements for storage of blood and blood related products in full compliance with 93/42/CE – class IIa regulation.
TSB Series Blood Bank Refrigerators are purpose built to meet the stringent guidelines to meet 93/42/CE class IIa regulation as a medical device for storage of blood and blood related products.
  • Wide range of sizes from 140L undercounter to 1500L double door.
  • Touchscreen digital controller with onboard datalogging
  • Whisper quite operation < 50 dBA
  • Triple pane glass doors.
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low energy consumption with real time energy monitoring
  • Low heat output
  • Rear access port
  • Electronic lock with password protection
TRUSTED_SUSTAINABILITY
Specifica
Descrizione TSB Series Blood Bank Refrigerator
Capacità (sistema metrico) 700 L
Gamma di temperatura 2°C to 8°C
Temperatura (punto di regolazione) +4°C
Refrigerante R290
No. of Doors 1
N. di cassetti 7
Capacità (sistema metrico) ripiano 30 kg
Materiale armadietto Steel
Interno Painted Steel
Certificazione/Conformità 93/42/CEE, 2014/30/UE, 2014/35/UE, 2014/53/UE (only if wireless option), 2011/65/UE
Ruote Yes
Tipo di controllo Touchscreen Control with Password Protection
Porte 1 Rear Access Port
Warranty 36 months
Sbrinamento Adaptive Defrost
Opzioni di monitoraggio Alarm
Compatibilità Smart View Smart-Vue Pro Compatible
Dimensioni interne (L x P x A) 686 x 600 x 1500 mm
Dimensioni esterne (L x P x A) 840 x 750 x 2085 mm
Tipo di spina EU
Requisiti elettrici 230 V/50 Hz
Frequenza 50 Hz
Peso di spedizione (sistema metrico) 215 kg