missing translation for 'onlineSavingsMsg'
Learn More

TSFM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11701-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898953

  • 121.38€ / 20µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).
Specifica

Specifica

TSFM
Polyclonal
Unconjugated
TSFM
COXPD3, EF TS, EF Tsmt, TSFM
Rabbit
Antigen Affinity Chromatography
RUO
10102
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P43897
TSFM
TSFM Fusion Protein Ag2308
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali