Learn More
Thermo Scientific™ TSG General Purpose Freezers - Crated
Codice prodotto. 17147758 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
400 L
700 L
1500 L
Tipo di spina:
EU
Dimensione della confezione:
Pezzo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Tipo di spina Dimensione della confezione
17147758 700 L EU Pezzo
17127748 400 L EU Pezzo
17107738 1500 L EU Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 17147758 Fornitore Thermo Scientific™ N. del fornitore TSG700FESCV
  • ONLINE EXCLUSIVE PRICE
    6418.43€
    6020.00€ / pezzi 1
    Risparmia 398.43€
    6% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 25-02-2027
The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
  • Wide range of sizes from solid door 400 L to 1500 L double door.
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY
Specifica
Descrizione TSG Series General Purpose Laboratory Freezer
Capacità (sistema metrico) 700 L
Gamma di temperatura -12°C to -30°C
Temperatura (punto di regolazione) -20°C
Door Style Solid
No. of Doors 1
Materiale armadietto Steel
Interno Painted Steel
Finitura esterna Painted Steel
Ripiani 4 Adjustable
Capacità (sistema metrico) ripiano 30 kg
Certificazione/Conformità CE
Ruote Yes
Tipo di controllo Digital Microprocessor
Porte 1 Rear Access Port
Warranty 12 Months
Sbrinamento Adaptive Defrost
Tipo di imballaggio Crated
Dimensioni interne (L x P x A) 686 x 600 x 1505 mm
Dimensioni esterne (L x P x A) 686 x 600 x 1505 mm
Opzioni di monitoraggio Alarm
Sicurezza dell'accesso Key Lock
Compatibilità Smart View Smart-Vue Pro Compatible
Tipo di spina EU
Tensione 230 V
Frequenza 50 Hz
Green Features Ad alta efficienza energetica
Refrigerante R290
Peso di spedizione (sistema metrico) 171 kg