missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPYL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551264
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total TSPYL1 protein.
TSPYL1 Polyclonal antibody specifically detects TSPYL1 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifica
TSPYL1 | |
Polyclonal | |
Unconjugated | |
TSPYL1 | |
testis-specific protein, Y-encoded-like 1; testis-specific Y-encoded-like protein 1; TSPY like 1; Tspyl; Tspyl1; TSPY-like 1; TSPY-like protein 1; Y encoded | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22110, 7259 | |
-20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O88852, Q9H0U9 | |
TSPYL1 | |
Synthetic peptide of human TSPYL1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |