Learn More
TSQ, TRC
Product Code. 30456872
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456872 50 mg 50mg
30450385 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456872 Supplier Toronto Research Chemicals Supplier No. TRCT94755350MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109628-27-5
Molecular Formula C17H16N2O3S
Molecular Weight (g/mol) 328.39
Synonym N-(6-Methoxy-8-quinolinyl)-4-methylbenzenesulfonamide, NSC 120213, TSQ
IUPAC Name N-(6-methoxyquinolin-8-yl)-4-methylbenzenesulfonamide
SMILES COc1cc(NS(=O)(=O)c2ccc(C)cc2)c3ncccc3c1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C17H16N2O3S/c1-12-5-7-15(8-6-12)23(20,21)19-16-11-14(22-2)10-13-4-3-9-18-17(13)16/h3-11,19H,1-2H3
Formula Weight 328.0882
Chemical Name or Material TSQ