Learn More
(+)-Tubocurarine Chloride Hydrochloride Pentahydrate, TRC
Specifica
Specifica
| Materiale o nome chimico | (+)-Tubocurarine chloride hydrochloride pentahydrate |
| CAS | 6989-98-6 |
| Formula molecolare | C37 H41 N2 O6 . Cl . Cl H . 5 H2 O |
| Quantità | 100 mg |
| Sinonimo | 13H-4,6:21,24-Dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium','2,3,13a,14,15,16,25,25a-octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-','chloride','hydrochloride','pentahydrate','(13aR,25aS)-, Tubocuraranium','7',12'-dihydroxy-6,6'-dimethoxy-2,2',2'-trimethyl-','chloride','hydrochloride','pentahydrate, (+)-Tubocurarine chloride hydrochloride pentahydrate, Tubarine pentahydrate, d-Tubocurarine dichloride pentahydrate |
| InChI Formula | InChI=1S/C37H40N2O6.2ClH.5H2O/c1-38-14-12-24-19-32(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-37-35-25(20-34(43-5)36(37)41)13-15-39(2,3)29(35)17-23-8-11-30(40)31(18-23)45-33;;;;;;;/h6-11,18-21,28-29H,12-17H2,1-5H3,(H-,40,41);2*1H;5*1H2/t28-,29+;;;;;;;/m0......./s1 |
| SMILES | O.O.O.O.O.Cl.[Cl-].COc1cc2CC[N+](C)(C)[C@@H]3Cc4ccc(O)c(Oc5cc6[C@H](Cc7ccc(Oc(c1O)c23)cc7)N(C)CCc6cc5OC)c4 |
| IUPAC Name | (13aR,25aS)-2,3,13a,14,15,16,25,25a-Octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-13H-4,6:21,24-dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium chloride hydrochloride pentahydrate |
| Molecular Weight (g/mol) | 771.72 |
| Peso formulazione | 770.2948 |