Learn More
WTW™ Turbidimeter Cuvette
Product Code. 11884602
Change view
Click to view available options
For Use With (Equipment):
Photoflex Series and Turb 430 IR/T Turbidimeter and Photometer
Turb 350/355 IR and 355 T Portable Turbidimeter
Turb 550, Turb 550 IR, Turb 555 and Turb 555 (IR) Laboratory Turbidimeter
Unit Size:
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11884602 Photoflex Series and Turb 430 IR/T Turbidimeter and Photometer Pack of 3
11786145 Turb 350/355 IR and 355 T Portable Turbidimeter Pack of 3
10181481 Turb 550, Turb 550 IR, Turb 555 and Turb 555 (IR) Laboratory Turbidimeter Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11884602 Supplier WTW™ Supplier No. 251302
 Restricted Item
In stock
For automatic detection of the photLab™ turbidimeters and spectral photometers
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
TRUSTED_SUSTAINABILITY
Specifications
Type Cuvette
For Use With (Equipment) Photoflex Series and Turb 430 IR/T Turbidimeter and Photometer
Quantity 3 Pack