Learn More
WTW™ Turbidity Standard
Product Code. 10478413
Change view
Click to view available options
Turbidity:
0.02, 10, 100,1750 NTU
10,000 NTU
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Turbidity Unit Size
10478413 0.02, 10, 100,1750 NTU Each
10134273 10,000 NTU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10478413 Supplier WTW™ Supplier No. 600543
  • ONLINE EXCLUSIVE PRICE
    596.66€
    520.00€ / pezzi 1
    Risparmia 76.66€
    12% Sconto
 Restricted Item
Spedizione stimata: 21-12-2026
Turbidity standards AMCO Clear™ with the highest precision and long-term stability are N.I.S.T. traceable, lot certified and not hazardous to your health.
  • Long-term precision and stability
  • No health hazards
  • Easy to dispose of
  • Adapted to the respective device optics in their particle composition
  • Suitable for applications in the lower measuring range such as drinking water
TRUSTED_SUSTAINABILITY
Specifications
Type Turbidity Standard
For Use With (Equipment) Turb 555 IR
Description Calibration standards
Includes Cleaning Tissue and Marking Rings
Turbidity 0.02, 10, 100,1750 NTU