missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turpentine Substitute, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantità:
1 L
2.5 L
25 L
Imballaggio:
Amber glass bottle
HDPE plastic bottle
Metal can
Plastic Coated Glass bottle
Specifica
Colore | Colorless |
Punti di ebollizione | 149°C |
Imballaggio | Plastic Coated Glass bottle |
Quantità | 1 L |
Forma fisica | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'