Learn More
Fryka™ TUS// Logg Underbench Freezer
Product Code. 12669345
Change view
Click to view available options
Capacity (Metric) Freezer:
100 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Freezer Unit Size
12669345 100 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12669345 Supplier Fryka™ Supplier No. 080TUS80100L
  • ONLINE EXCLUSIVE PRICE
    15700.23€
    12181.98€ / pezzi 1
    Risparmia 3518.25€
    22% Sconto
 Restricted Item
Spedizione stimata: 26-02-2027
Fryka TUS// Logg Underbench Freezer has usable space of 100L and can be used for cooling and freezing in industrial and scientific laboratories.
  • Colour touch screen controller with representation of the temperature profile and user friendly menu structure
  • Integrated data logger readable over USB stick for temperature profile and alarm events
  • Battery-supplied alarm, Ethernet interface // in case of an alarm event E-Mail will be sent
  • Automatic and detailed supervision function of refrigerating machine with failure analysis
  • Remote maintenance with worldwide access over internet
  • Detailed user administration to protect adjusted alarm- and temperature set points
  • Registered access control
  • Vacuum-Insulation: large interior space with small external dimensions and low power consumption
  • All stainless steel casing
  • Bushing (diameter 19mm), e.g. for temperature monitoring with external sensors
  • Refrigeration unit: low noise, fully hermetically sealed, air-cooled, low maintenance
  • TUS 80-100: two-stage cascade refrigeration system
  • Foamed door with double door seal
  • Air vents at front and right, making it suitable for installation under tables
  • Potential-free contact for connection to external alarm system
  • 100mV/K-output for connection to temperature recording system
  • Lockable door
  • With inlaid grilles and four shelf rails
  • Stackable unit, max. 2 units
Specifications
Capacity (Metric) Freezer 100 L
Temperature Range (Refrigerator) 50°C to 80°C
Style Undercounter
For Use With (Application) Industrial and scientific laboratories
Includes Integrated data logger and touch screen controller
Model TUS 80100 //logg
Dimensions (D x W x H) Interior 45 x 45 x 50 cm
Depth (Metric) Exterior 73 cm
Height (Metric) Exterior 75 cm
Width (Metric) Exterior 95 cm
Electrical Requirements 230 V 50 Hz