Learn More
Lenz Laborglasintrumente™ Twisselmann Extractor Head
Product Code. 10692852
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10692852 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10692852 Supplier Lenz Laborglasintrumente™ Supplier No. 05415125
  • ONLINE EXCLUSIVE PRICE
    175.00€
    115.03€ / pezzi 1
    Risparmia 59.97€
    34% Sconto
Spedizione stimata: 23-12-2026
Twisselmann extractor head with vacuum jacket
  • With vacuum jacket
  • Made of DURAN™ tubing
Specifications
Product Type Condenser NS 29/32
Capacity (Metric) 30 mL
Material DURAN Tubing
Includes Vacuum jacket
Quantity 1