missing translation for 'onlineSavingsMsg'
Learn More

TXNIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18243-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898304

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.
Specifica

Specifica

TXNIP
Polyclonal
Unconjugated
TXNIP
EST01027, HHCPA78, THIF, Thioredoxin binding protein 2, TXNIP, VDUP1
Rabbit
Antigen Affinity Chromatography
RUO
10628, 56338
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BG60, Q9H3M7
Txnip
TXNIP Fusion Protein Ag13009
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali