Learn More
Masterflex™ Tygon™ E-3603 Non-DEHP Vacuum Tubing
Product Code. 15191323 Shop All Masterflex Products
Change view
Click to view available options
Diameter (Metric) Inner:
6.4 mm
9.6 mm
12.8 mm
Unit Size:
3m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
15191323 9.6 mm 3m
15137134 12.8 mm 3m
15147134 6.4 mm 3m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15191323 Supplier Masterflex™ Supplier No. 0641430
  • ONLINE EXCLUSIVE PRICE
    356.00€
    272.46€ / 3m
    Risparmia 83.54€
    23% Sconto
Spedizione stimata: 04-01-2027
Bends into sharp radius curves and easily works with fittings. Saint Gobain Performance Plastics™ Tygon™ E-3603 Non-DEHP Vacuum Tubing is for use with general laboratory, food and beverage, biopharmaceutical and analytical instruments.
Transparent tubing is Non-DEHP and phthalate free. Biodegradable crack-resistant and non-oxidizing.
  • Meets USP Class VI and NSF 51 requirements
  • Temperature range: –46 to 74°C
  • Vacuum service of 760mm Hg at 23°C
  • Sterilize: Autoclave or ethylene oxide
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 3/8 in.
Diameter (Metric) Inner 9.6 mm
Material Tygon
Diameter (English) Outer 7/8 in.
Overall Length (English) 10 ft.
Overall Length (Metric) 300 cm
Thickness (English) 0.25 in.
Thickness (Metric) 6.4 mm
For Use With For 10mm tubulation
For Use With (Application) General lab, food and beverage and biopharm.
Min. Operating Temperature (Metric) -46°C
Color Transparent
Maximum Pressure 47 psi
Vacuum Level 29.9 in. Hg
Max. Operating Temperature (Metric) 121°C
Product Type Tubing