Learn More
DuPont™ Tyvek 500 Laboratory Coats press studs
Product Code. 11762594 Shop All DuPont Products
Change view
Click to view available options
Size: Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11762594 Size: Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11762594 Supplier DuPont™ Supplier No. D13396079
  • ONLINE EXCLUSIVE PRICE
    401.83€
    305.09€ / pezzi 50
    Risparmia 96.74€
    24% Sconto
Spedizione stimata: 07-12-2026
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Collar provide an excellent barrier against fine particles and fibres, is ultra-low-linting and antistatically treated.
  • Silicon non-added
  • Stitched internal seams
  • 3 pockets and 5 press stud closures
  • Composed of flash spun high density polyethylene
  • Provides an idea balance of protection, durability and comfort
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
The user should make the risk assessment to determine the protection required.