missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ U-Cap Sport Bump Cap - Long Brim

An innovative bump cap with a hard shell element and honeycomb structure to absorb shock, in addition to greatly reducing the risk of head injuries. Long brim design provides increased sun protection.

Marca:  Uvex™ 9794408

Dettagli aggiuntivi : Peso : 0.22200kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17299221

  • 28.30€ / Pezzo

Spedizione stimata: 09-03-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Ideal for areas where helmets are not compulsory, but there is still a risk of head injuries
  • Innovative hard shell element in armadillo-design, honeycomb structure, and shock-absorbing elements
  • Reliable impact protection in accordance with EN 812
  • Long brim provides increased sun protection
Specifica

Specifica

Bump Cap
No
Long Brim
60 to 63 cm
EN 812:2012
Navy Blue
Armadillo-Style Hard Shell
9024 U-Cap Sport
Non-sterile
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali