Learn More
Uvex™ U-Chem 3300 Chemical Protection Gloves
Catalog Number 17280521
missing translation for 'orderingAttributeHoverText'
Size:
7
8
9
10
11
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
NBR-coated chemical protection gloves feature seamless, bamboo-viscose support material that is particularly kind to sensitive skin. Natural fibers help absorb moisture to prevent excessive perspiration.
  • NBR-coated chemical protection gloves with bamboo-fiber supporting material
  • Natural fibers are especially kind to sensitive skin
  • Comfortable to wear for extended periods of time
  • Resistant to a wide variety of chemicals
  • Silicone-free according to imprint test
Applicazioni Laboratory
Coating Material NBR
Colore Blue
Size 8
EN388 Standard 2121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
Da utilizzare con (applicazione) Chemical Handling, Food Processing, Inspection, Laboratory, Maintenance, and Pharmaceuticals
Sesso Unisex
Stile polso guanto Gauntlet
Materiale rivestimento interno Bamboo, Nylon
Serie U-Chem 3300
Lunghezza (sistema metrico) 32 cm
Afficher plus Afficher moins