missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567550
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
UBE1L Polyclonal antibody specifically detects UBE1L in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifica
UBE1L | |
Polyclonal | |
Unconjugated | |
UBA7 | |
1300004C08Rik; D8; MGC12713; UBA1, ubiquitin-activating enzyme E1 homolog B; UBA1B; UBA7; UBA7, ubiquitin-activating enzyme E1; UBE1L; UBE2; ubiquitin like modifier activating enzyme 7; ubiquitin-activating enzyme 7; Ubiquitin-activating enzyme E1 homolog; ubiquitin-activating enzyme E1-related protein; ubiquitin-activating enzyme-2; ubiquitin-like modifier activating enzyme 7; ubiquitin-like modifier-activating enzyme 7 | |
Rabbit | |
affinity chromatography | |
RUO | |
7318 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P41226 | |
UBA7 | |
Fusion protein of human UBE1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |