missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17056-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.Specifica
UBE2F | |
Polyclonal | |
Unconjugated | |
UBE2F | |
NCE2, NEDD8 carrier protein UBE2F, NEDD8 conjugating enzyme 2, NEDD8 protein ligase UBE2F, UBE2F | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140739, 363284, 67921 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5U203, Q969M7, Q9CY34 | |
Ube2f | |
UBE2F Fusion Protein Ag10764 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |