missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ UBE2R2 Recombinant Protein
Marca: Abnova™ H00054926-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
AAH47584 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
52.3 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MAQQQMTSSQKALMLELKSLQEEPVEGFRITLVDESDLYNWEVAIFGPPNTLYEGGYFKAHIKFPIDYPYSPPTFRFLTKMWHPNIYENGDVCISILHPPVDDPQSGELPSERWNPTQNVRTILLSVISLLNEPNTFSPANVDASVMFRKWRDSKGKDKEYAEIIRKQVSATKAEAEKDGVKVPTTLAEYCIKTKVPSNDNSSDLLYDDLYDDDIDDEDEEEEDADCYDDDDSGNEES | |
CDC34B/FLJ20419/MGC10481/UBC3B | |
UBE2R2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
54926 | |
UBE2R2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UBE2R2 | |
Human | |
Recombinant | |
Solution |