Learn More
CleanSpace™ Ultra Power System
Product Code. 15930102 Shop All CleanSpace Technology Products
Change view
Click to view available options
Power Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15930102 Power Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15930102 Supplier CleanSpace™ Supplier No. PAF0070
  • ONLINE EXCLUSIVE PRICE
    1865.00€
    1291.32€ / pezzi 1
    Risparmia 573.68€
    30% Sconto
 Restricted Item
Spedizione stimata: 17-12-2026
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Battery-powered, positive pressure air-purifying respirator with an IP Rating 66. Suitable for decontamination procedures and applications where there is an abundance of dust and water. Can be used for particulate and gas applications.
  • IP Rating 66 – resistant to dust and liquid ingress
  • Suitable for decontamination showers
  • Light weight and comfortable
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Long operating time: up to 8 hours and quick battery recharging: < 2 hours
  • Easy and accurate flow capability check
Specifications
Product Type PAPR Assembly, No Headtop
Model PAF-0070
Mask sold separately