Learn More
IKA™ ULTRA-TURRAX™ T 18 Digital Disperser
Product Code. 13396299 Shop All IKA Products
Change view
Click to view available options
Product Type:
Digital Disperser
Digital Disperser Package
Plug Type:
EU Plug
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Plug Type Unit Size
13396299 Digital Disperser EU Plug Each
13470657 Digital Disperser Package EU Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13396299 Supplier IKA™ Supplier No. 0003720000
  • ONLINE EXCLUSIVE PRICE
    1830.00€
    1738.22€ / pezzi 1
    Risparmia 91.78€
    5% Sconto
In stock
Disperses volumes from 1 to 1500mL (H2O) with digital speed display. IKA™ ULTRA-TURRAX™ T 18 Digital Disperser offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Competitively priced dispersing instrument for volumes from 1 to 1500mL (H2O) with digital speed display. Offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
  • Quiet operation
  • Motor protection against overload
  • Digital display for precise monitoring of set and actual speeds 2+1 year after registering at 2+1 www.ika.com/register
  • Rotating knob for adjusting the speed
TRUSTED_SUSTAINABILITY
Specifications
Speed 3000 to 25,000 rpm
Voltage 220/240 V
Electrical Requirements 220/240 V, 50/60 Hz, 500 W
Wattage 500 W
Volume Range (Metric) 1 mL to 1.5 L
Depth (Metric) 106 mm
Height (Metric) 450 mm
Width (Metric) 87 mm
Display LED
Includes Electronic Speed Control and Overload Protection. With EU plug.
Model T 18 ULTRA-TURRAX™
Packaging Quantity 1 Ea.
Plug Type EU Plug
Viscosity 5000 mPas
Product Type Digital Disperser
Frequency 50 Hz