Descrizione
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast GC columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific TRACE GC UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
Applications:
Chemical
Petrochemical
Environmental
Flavors and fragrances
UltraFast GC columns utilize short (2 to 10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
Dramatically shorter analysis times
Increased sample throughput–by a factor of 20
Longer column lifetimes
UltraFast UFC-WAX Columns uses:
Fatty acid methyl esters (FAMEs)
Essential oils
General
Specifications
Specifications
| Da utilizzare con (applicazione) | GC for Chemicals, GC for Petrochemicals, GC for Environmental Analysis, GC for Flavors and Fragrances |
| Lunghezza (sistema metrico) | 5 m |
| Diametro (sistema metrico) interno | 0.1 mm |
| Stationary Phase | UltraFast UFC-WAX |
| Da utilizzare con (apparecchiatura) | Trace GC UltraFast Instrument |
| Descrizione | 5 m x 0.1 mm x 0.1 μm |
| Product Line | Ultrafast |
| Spessore pellicola | 0.1 μm |