Learn More
Beskrivelse
These flexible goggles can also be worn as overspecs with prescription spectacles and are available with cellulose acetate or polycarbonate lenses, as welding safety spectacles or with a flip-up face guard.
General features
- Classic safety goggle suitable for a wide range of applications
- Indirect ventilation system increases air circulation
- A sustainable benefit: lenses can be replaced when required
- Available replacement lens: 9300956
- Metal-free
Safety features
- Certified in line with EN 166 (personal protective eyewear) and EN 170 (UV protective filter)
- Marking: W 166 3 F CE – 2C-1,2 W1 FN CE
- Cellulose acetate lens with anti-fog performance on the inside and UV380 protection
Comfort features
- Flexible face seal naturally moulds to any facial shape
- Generous internal volume accommodating prescription spectacles and frame recesses to locate the side arms ensuring a comfortable fit
- Length-adjustable textile headband for a secure fit and individual adaptability
Tekniske data
Tekniske data
| Tipo di lente | Chemical-resistant |
| Lens Coating | Anti-fog |
| Colore | Transparent Gray |
| Certifications/Compliance (Europe) | EN 166, EN 170 |
| Lens Tint | Clear |
| Product Type | Anti-fog Goggles |
| Materiale fascia per la testa | Textile |
| Materiale | Polycarbonate |
| Monouso | Reusable |
| Lens Material | Cellulose Acetate |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?