Learn More
UNC45A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558703
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: QELQHRGAVV VLNMVEASRE IASTLMESEM MEILSVLAKG DHSPVTRAAA ACLDKAVEYG LIQPNQDG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.
UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen.Specifica
UNC45A | |
Polyclonal | |
Unconjugated | |
UNC45A | |
AW538196; GCUNC45; GC-UNC45; GCUNC-45; general cell UNC45; IRO039700; Protein unc-45 homolog A; RGD1305357; SMAP1; SMAP-1; smooth muscle cell associated protein-1; smooth muscle cell-associated protein 1; Stromal membrane-associated protein 1; unc-45 homolog A; unc-45 homolog A (C. elegans); unc-45 myosin chaperone A; Unc45a; UNC-45A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55898 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3U1 | |
UNC45A | |
Recombinant protein corresponding to Human UNC45A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only