missing translation for 'onlineSavingsMsg'
Learn More

Undecanophenone, 97%, Thermo Scientific™

Product Code. 11311517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311517 5 g 5g
11321517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311517 Supplier Thermo Scientific Alfa Aesar Supplier No. L02119.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4433-30-1
Molecular Formula C17H26O
Molecular Weight (g/mol) 246.394
MDL Number MFCD00051548
InChI Key LHJBFOGCFZHBAJ-UHFFFAOYSA-N
Synonym undecanophenone, n-undecanophenone, 1-phenylundecanone, 1-undecanone, 1-phenyl, decyl phenyl ketone, 1-phenyl-1-undecanone, 1-phenyl-undecan-1-one, 1-phenyl-1-undecanone #, acmc-209jx6
PubChem CID 78167
IUPAC Name 1-phenylundecan-1-one
SMILES CCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 28°C to 30°C
Boiling Point 202°C to 204°C (14 mmHg)
Quantity 5 g
Beilstein 2211502
Formula Weight 246.39
Percent Purity 97%
Chemical Name or Material Undecanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.