Learn More
Sartorius™ Replacement Screen Kits for Use with Glass Holders for Vacuum Filtration Membranes
Sfoglia Tutto Sartorius ProdottiDescrizione
- Product Type: Replacement Screen Kits
- Material: PTFE (Main Ring), PTFE/Stainless Steel (Drain Screen, Support Screen), Silicone (O-Ring)
- Includes: Support Screen, Drain Screen, Main Ring, O-Ring
- For Use With (Equipment): Sartorius™ Glass Holders for Vacuum Filtration Membranes
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Filter Holder |
| Materiale | Politetrafluoroetilene (PTFE), Stainless Steel, Silicone |
| Sterilità | Non-sterile |
| Da utilizzare con (applicazione) | Vacuum Filtration |
| Capacità | Series 16316 Glass Holders for Vacuum Filtration Membranes |
| Include | Main Ring, Drain Screen, Support Screen, O-Ring |
| Modello | 17147 |
| Tipo | Screen Kit |
| Diametro (sistema metrico) membrana | 50 mm/47 mm |