Learn More
Abeba™ UNI6 31753 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Marca:  Abeba™ 31753/48
Dettagli aggiuntivi : Peso : 1.16000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15619137
  • 89.85€ / 1 paio
Spedizione stimata
19-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 S2 SRC
  • Smooth leather
  • Black in ATEX design
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Discontinued model
  • **Size 35 with replaceable acc Wave insole (art. 3576)
Specifica
Specifica
CE EN ISO 20345:2011 S2 SRC
Microfiber
48
Polyurethane
Steel
Black
Leather
Slip-resistant
31753