Learn More
Abeba™ UNI6 31763 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Marca:  Abeba™ 31763/42
Dettagli aggiuntivi : Peso : 1.14000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15619187
  • 86.20€ / 1 paio
Spedizione stimata
24-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 S1 SRC
  • Functional leather
  • Dark blue
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • **Size 35 with replaceable acc Wave insole (art. 3576)
  • Water-repellent
Specifica
Specifica
CE EN ISO 20345:2011 S1 SRC
Leather
Slip-resistant
31763
Blue
42
Polyurethane
Steel