Learn More
Abeba™ UNI6 36780 Shoes
Descrizione
- CE EN ISO 20347:2012 O2 FO SRC
- Microfiber
- White
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 O2 FO SRC |
| Colore | White |
| Materiale | Microfiber |
| Size | 37 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 36780 |