Learn More
ABEBA™ Uni6 Anthracite Safety Trainers S2 ESD
Descrizione
- Made for comfort and safety
- Comply to EN ISO 20345: 2012, S2, SRC
- ESD dissipative according EN 61340
- Slip resistant PU/TPU outsole
- Breathable and water repellent knitted fabric
- Steel toe cap (200 J)
- Breathable and antibacterial lining with silver ions
- Removable insole active comfort
Specifica
Specifica
| Colore | Anthracite |
| Materiale | Knitted Fabric |
| Size | 47 |
| Materiale dello zoccolo | Steel |
| Certifications/Compliance (Europe) | ESD according 61340 class climatic 1, EN ISO 20345: 2012 S2 SRC |
| Sesso | Unisex |
| Ankle Style | Trainers |
| Waterproof | Water Repellent |
| Product Type | S2 SRC ESD |