Descrizione
- Gloves feature two-layer material composition of acrylic and new wool
- Polymer coating on the palm remains flexible even at low temperatures
- Coating provides mechanical abrasion resistance
- Enhanced grip capabilities in dry, or slightly damp areas
- Provides thermal insulation when in direct contact with cold objects
Specifica
Specifica
| Coating Material | Polymer |
| Colore | Black |
| Finitura esterna | Smooth |
| Hand | Ambidestri |
| Materiale | Polyamide, Elastane |
| Product Type | Cold-resistant Gloves |
| Size | 9 |
| EN388 Standard | 3131X |
| Stile polso guanto | Knit |
| Materiale rivestimento interno | Acrylic, Wool |
| Vedi altri risultati |