Learn More
Simport™ Scientific Unisette™ Biopsy Processing/Embedding Cassette in Quickload™ stacks for Primera printers
Secure specimens during processing with Simport Scientific™ Unisette™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
Marca: Simport™ Scientific M406-3T
Dettagli aggiuntivi : Peso : 3.91500kg
Descrizione
In Quickload™ stacks, for Primera printers
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevent early separation of base and lid
- Snap latch and hinge-lock design - cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 25 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifica
1000 | |
Pink | |
Acetal |