Learn More
Simport™ Scientific Unisette™ Biopsy Processing/Embedding Cassette in Quickload™ stacks for Primera printers
Secure specimens during processing with Simport Scientific™ Unisette™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
Marca:  Simport™ Scientific M406-3T
Dettagli aggiuntivi : Peso : 3.91500kg
 vedi altre versioni di questo prodotto
Codice prodotto. 16594921
  • 167.00€ / pezzi 1000
Spedizione stimata
22-04-2021
Aggiungi al carrello
Descrizione
Descrizione
In Quickload™ stacks, for Primera printers
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 25 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifica
Specifica
1000
Pink
Acetal