Learn More
S Murray and Co™ Universal Bandage Scissors
Product Code. 12962065
Change view
Click to view available options
Length (Metric):
175 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12962065 175 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12962065 Supplier S Murray and Co™ Supplier No. D917/05
  • ONLINE EXCLUSIVE PRICE
    14.95€
    12.81€ / pezzi 1
    Risparmia 2.14€
    14% Sconto
Spedizione stimata: 23-12-2026
Use these scissors for cutting anything from bandages to stainless steel mesh. S Murray™ Universal Bandage Scissors are heavy duty but lightweight with stainless steel blades and robust construction.
  • Nickel plated steel
  • Straight with one sharp point
  • For cutting anything from bandages to stainless steel mesh
  • Autoclavable
Specifications
Product Type Universal Bandage Scissors
Blade Style Straight
Length (Metric) 175 mm
Material Stainless Steel
Autoclavable Autoclavable