Learn More
JULABO™ Universal Basic Tray for Assembling Spring Clamps
For assembling Erlenmeyer flasks from 25 to 1000mL, icluding spring clamps. This accessory is suitable for JULABO™ SW22, SW23.
Marca: JULABO™ 8970630
Dettagli aggiuntivi : Peso : 0.25000kg
Specifica
SW22, SW23 shaking water bath | |
Universal Basic Tray | |
25 to 1000 mL Erlenmeyer Flasks | |
Spring clamp |