Learn More
Pacific Handy Cutter Universal Nylon Utility Holster with Pen Loop
Product Code. 17759047
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17759047 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17759047 Supplier Pacific Handy Cutter Supplier No. UKH671
  • ONLINE EXCLUSIVE PRICE
    220.46€
    185.85€ / pezzi 10
    Risparmia 34.61€
    15% Sconto
Spedizione stimata: 24-12-2026
Practical belt holster made of nylon with loop for belt attachment.
  • The holster also has a convenient pen slot.
  • Suitable for most knives, especially QBA375 and QBS20.
Specifications
Product Type Holster
For Use With (Equipment) Most Knives, Especially QBA375 and QBS20
Material Nylon
Color Black
No. per Pack 10/Pk.
Sterility Non-sterile