Learn More
Kinesis™ Quick Connect Adapter
Product Code. 12967769
Change view
Click to view available options
Type:
Female Luer Adapter
Male Luer Adapter
Material:
ETFE
PEEK
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material Unit Size
12967769 Male Luer Adapter ETFE Each
12937759 Female Luer Adapter PEEK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12967769 Supplier Kinesis™ Supplier No. 0201421
  • ONLINE EXCLUSIVE PRICE
    20.85€
    19.99€ / pezzi 1
    Risparmia 0.86€
    4% Sconto
Spedizione stimata: 03-12-2026
Add to basket
Add to basket
Convenience of quick connect and disconnect of a luer connection
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Specifications
Type Male Luer Adapter
Diameter (English) Inner 0.05 in.
Temperature (Metric) Max 40°C
Material ETFE
Color Red
For Use With (Equipment) Male Luer Lock x Female 1/4-28 Flat Bottom