Learn More
Liebherr™ Upright Pharmacy Fridge with Shelves
Product Code. 17889096 Shop All Liebherr Appliances Products
Change view
Click to view available options
Capacity (Metric):
394 L
420 L
558 L
Door Style:
Glass Self-closing
Solid Reversible Self-closing with Lock
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Door Style Unit Size
17889096 420 L Glass Self-closing Each
17819106 394 L Solid Reversible Self-closing with Lock Each
17818306 558 L Solid Reversible Self-closing with Lock Each
17808306 558 L Glass Self-closing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17889096 Supplier Liebherr™ Supplier No. HMFVH4011
  • ONLINE EXCLUSIVE PRICE
    3530.00€
    3390.00€ / pezzi 1
    Risparmia 140.00€
    3% Sconto
 Restricted Item
Spedizione stimata: 11-12-2026
Secure, accurate and ultra energy-efficient fridge storage of temperature-sensitive pharmacy products, the Liebherr™ Pharmacy Fridge is fully-equipped for pharmacy applications with Liebherr build-quality you can rely on.
  • Upright pharmacy fridge, reach-in, 1 section, DIN 13277 compliant
  • Temperature set to 5°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration
  • Refrigerant R 600a, connection rating 2.0 A, 220-240V/50/1-ph,ultra-efficient
  • Digital controller with 2.4 in. touch and swipe color display and 0.1°C resolution, password keypad lock
  • Connectivity: Integrated LAN/WLAN interface and integrated data logger with USB-port
  • Access port for external temperature probe, additional safety thermostat
  • Advanced optical alarms and alarm history with min/max temperatures, duration etc.
  • Interior lighting: vertical LED on left-side wall
  • Electronic door lock with remote control, reversible hinges, self-closing door with
  • 5-year extended warranty option available in select countries
Specifications
Type Pharmacy Fridge
Capacity (Metric) 420 L
Temperature Range 5°C
Temperature Sensor PT1000
Refrigerant R 600a
Door Style Glass Self-closing
No. of Doors 1
Shelves 5
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish White Powder-coated
Certifications/Compliance ISO 9001, ISO 14001, CE
Style Upright
Display 2.4 in Touch and Swipe Color Display in Door
Casters No
Color White
Condenser Type Low-maintenance Rear-wall Wire-on-tube
Ports 1 x 10 mm dia.
Warranty 2 Years Parts and Labor, 5 Year Optional Extended Warranty
Defrost Timed Off-cycle
Model HMFvh 4011
Monitoring Options SmartCoolingHub
Compressor CFC-free
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Phase 1, Single Phase