Learn More
WTW™ USB Adapter for WTW™ Meters
Product Code. 11881603
Change view
Click to view available options
For Use With (Equipment):
Laboratory instruments (except OxiCal OC 100/110)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11881603 Laboratory instruments (except OxiCal OC 100/110) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11881603 Supplier WTW™ Supplier No. 902881
Spedizione stimata: 29-12-2026
The USB adapters ADA USB/Ser and ADA USB are used to connect devices with RS 232 interface
WTW offers adapter solutions for mobile energy supply, data transfer of old systems as well as for the connection of conventional pH electrodes to digital systems. The USB adapters ADA USB/Ser and ADA USB are used to connect devices with RS 232 interface to PCs and laptops with USB interfaces, especially all lab and portable meters with RS 232 connector (portable meters 3xxi, benchtop meters inoLab™ Level 2, 730, field meters 197i, 1970i).
TRUSTED_SUSTAINABILITY
Specifications
Description USB Adapter
For Use With (Equipment) Laboratory instruments (except OxiCal OC 100/110)