Learn More
Grant Instruments™ UV Cabinet
Codice prodotto. 13424356
Cambia vista
Click to view available options
Portata:
7 m3/h
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Portata Unit Size
13424356 7 m3/h Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13424356 Supplier Grant Instruments™ Supplier No. UVTBAR EURO
  • ONLINE EXCLUSIVE PRICE
    2795.00€
    2467.55€ / pezzi 1
    Risparmia 327.45€
    11% Sconto
View all promotions
 Articolo soggetto a restrizioni
Spedizione stimata: 29-12-2026
UVT-B-AR Economy bench-top model for protection against contamination during a variety of DNA/RNA procedures, with dual UV lamp protection.
  • UV surface irradiation – via single 25W 254nm open UV lamp
  • Patented high intensity UV air cleaner – 25 m3/hour cleaner recirculator continuous air flow with 1cm UV irradiation distance
  • UV exposure control – 24 hour digital timer
  • Built-in power socket
  • UV protection - UV protective film on glass panels
  • Convenient, easy to use digital timer for accurate control of UV exposure
  • White lamp provides local illumination of the workplace to optimise visual control during operations
  • Shock proof glass front, stainless steel sides, metal framework and stainless steel work surface
  • Patented built-in UV cleaner recirculator prevents unwanted contamination and protects the user from direct UV light during manipulation
  • Second UV lamp disinfects the working area, inactivating DNA/RNA fragments during 15-30 min of exposure. Automatic switch-off when door is opened
  • Contains an integral power socket
TRUSTED_SUSTAINABILITY
Specifications
Product Type UV Cabinet
Tipo di illuminazione 1 open UV lamp 25 W bactericidal, 254 nm, ozone free, 1 UV recirculator, 25 W (efficiency >99% per 1 cycle), 1 white lamp
Materiale Stainless Steel, Glass
Amperaggio 0.3 A
Profondità (sistema metrico) 585 mm
Depth (Metric) Interior 475 mm
Portata 7 m3/h
Altezza (sistema metrico) 555 mm
Include Bactericidal air recirculator, 25m3/h air flow exchange
Tensione 120/230 V
Larghezza (sistema metrico) 690 mm
Larghezza (sistema metrico) interno 650 mm
Frequenza 50/60 Hz