Learn More
Grant Instruments™ UVC/T-M-AR EURO - General Purpose Stainless Steel PCR UV Cabinet
Codice prodotto. 16637865
Cambia vista
Click to view available options
Portata:
25 m3/h
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Portata Dimensione della confezione
16637865 25 m3/h Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16637865 Fornitore Grant Instruments™ N. del fornitore UVC/TMAR EURO
  • ONLINE EXCLUSIVE PRICE
    3305.00€
    2831.56€ / pezzi 1
    Risparmia 473.44€
    14% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 21-12-2026
Aggiungi al carrello
Aggiungi al carrello
UVC/T-M-AR EURO is an efficient general purpose stainless steel PCR UV cabinet workstation for carrying out short-term procedures.
The Grant Instruments UVC/T-M-AR EURO is an efficient general purpose stainless steel PCR UV cabinet workstation for carrying out short-term procedures.
The UVC/T-M-AR EURO cabinets are equipped with an open UV lamp installed in the upper part of the hood. UV-radiation from these lamps effectively disinfects the working area chamber, inactivating DNA / RNA fragments during 15-30 minutes of exposure. The digital timer controls the duration of the direct UV irradiation. An additional daylight lamp provides a bright, clear working area.
Models are equipped with a high efficiency flow-type bactericidal UV cleaner-recirculator which provide constant decontamination inside the cabinet during operation. The UV cleaner-recirculator consists of UV lamp, dust filters and a fan. The recirculation effect increases the maximum density of the UV light making it sufficiently effective for DNA / RNA inactivation. The recirculator processes 360 UV-cabinet volumes per hour, creating permanently aseptic conditions within the cabinet.
Designed for robustness and ease of access, it features a built-in UV cleaner-recirculator to prevent unwanted contamination and to protect the user from direct UV light during manipulation.
  • Built-in patented UV cleaner-recirculator both increases the maximum density of UV light and generates 25 m³/h air flow exchange – prevents unwanted contamination and protects the user from direct UV light during manipulation
  • UV surface irradiation – via single 25 W 254 nm open UV lamp
  • High intensity UV cleaner – 25 m³/hour cleaner-recirculator continuous air flow with 1 cm UV irradiation distance
  • UV protection – UV-protective film on glass panels
  • UV exposure controlled by 24h digital timer
  • Convenient, easy to use digital timer for accurate control of UV exposure
  • White lamp provides local illumination of the workplace to optimise visual control during operations
  • Irradiated UV light automatic switch-off when door opened
  • Front opening with three adjustable positions for ease of access
  • Suppled with 1 access port and internal power socket
Applications:
These PCR UV-cabinets are designed for clean operations with DNA samples and provide protection against contamination. They are not recommended for operations with dangerous infectious and viral materials as well as DNA / RNA amplification.
Specifications:
  • Dimensions (h x d x w): 535 x 515 x 690 mm
  • Construction: Stainless steel frame and working area
  • Panels: Glass with UV-protective film
  • Front opening with three adjustable positions : •
  • Open UV lamp, 25 W bactericidal, 254 nm, ozone free: 1
  • Bactericidal air recirculator, 25 m³/h air flow exchange: •
  • UV recirculator, 25 W (efficiency >99% per 1 cycle): 1
  • Day light white lamp for workplace illumination 15 W: 1
  • Radiation type: Ultraviolet (253.7 nm), ozone free
  • Optical transmission: 95%
  • Digital timer (non stop) 1 min – 24 hours: •
  • Internal power outlets: 1
  • Internal working area: 645 x 490 mm
  • Flow rate: 7
  • Power consumption: 67 W (0.3 A)
  • Nominal operating voltage: 120 or 240 V (50/60 Hz)
  • Weight (net): 28.8 kg
TRUSTED_SUSTAINABILITY
Specifica
Dimensioni (L x P x A) 515 x 690 x 535 mm
Portata 25 m3/h
Lunghezza (sistema metrico) interno 645 mm
Larghezza (sistema metrico) interno 490 mm