Learn More
uvex C300 Foam Cut Protection Gloves
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
7
8
9
10
Dimensione della confezione:
Pezzo
Descrizione
- Gloves provide level 3/C blade cut resistance, abrasion resistance, and high tear resistance
- High-performance elastomer (HPE) coating on the palm and fingers for enhanced grip capabilities
- Soft-grip foam coating provides mechanical abrasion resistance
- Highly flexible with very good tactile sensation
- Uvex Climazone technology reduces excessive moisture and warmth
- 3D Ergo technology for a secure fit
- Silicone-free according to the imprint test
Specifica
Specifica
| Area rivestita | Palm and fingertips |
| Coating Material | HPE |
| Colore | Gray |
| Cut Resistance Rating | ANSI A3 |
| Hand | Ambidestri |
| Latex Free | Sì |
| Materiale | HPPE, Polyamide, Glass, Bamboo Viscose |
| Product Type | Cut-resistant Gloves |
| Size | 10 |
| EN388 Standard | 3X42C |
| Vedi altri risultati |