Descrizione
- Wraparound style spectacle with excellent field of vision
- Straight side arms grip the side of the head not behind the ears. Provides a secure comfortable, slip free wear
- Combines hard/soft components - increased comfort around sensitive nose, brow and ear areas
- Fully adjustable side arms enables wearer to achieve individual fit for better protection and comfort
- 100% UV protection and metal free
Specifica
Specifica
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Protective Eyewear I-vo 9160 |
| Colore telaio | Blue, Gray |
| N. per confezione | 1 ciasc. |
| Materiale | Polycarbonate |
| Monouso | Reusable |
| Lens Material | Policarbonato |
| Materiale del telaio | Plastic |
| UV Protection | UV400 |
| Vedi altri risultati |