Learn More
uvex Occhiali protettivi I-vo 9160 Uvex™
Product Code. 11249869 Shop All Uvex Products
Change view
Click to view available options
Colore telaio:
Black, Gray
Blue, Gray
Blue, Orange
Nero/Grigio
Lens Tint:
Brown
Clear
Gray
Trasparente
Unit Size:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Colore telaio Lens Tint Unit Size
11249869 Blue, Orange Brown Pezzo
10345772 Black, Gray Gray Pezzo
10031072 Nero/Grigio Trasparente Pezzo
10777914 Blue, Gray Clear Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11249869 Supplier uvex Supplier No. 9160068
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Wraparound style spectacle with excellent field of vision
  • Wraparound style spectacle with excellent field of vision
  • Straight side arms grip the side of the head not behind the ears. Provides a secure comfortable, slip free wear
  • Combines hard/soft components - increased comfort around sensitive nose, brow and ear areas
  • Fully adjustable side arms enables wearer to achieve individual fit for better protection and comfort
  • 100% UV protection and metal free
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Brown
Product Type Protective Eyewear I-vo 9160
Colore telaio Blue, Orange
N. per confezione 1 ciasc.
Materiale Polycarbonate
Monouso Reusable
Lens Material Policarbonato
Materiale del telaio Plastic
UV Protection UV400
Frame Type Standard