Learn More
Cole-Parmer™ V Series Stuart™ Vortex Mixer
Product Code. 12017134
Change view
Click to view available options
Speed Range:
200 to 2500 rpm
2500 to 4500 rpm
Fixed Speed
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
12017134 Fixed Speed Each
12027134 200 to 2500 rpm Each
15401435 2500 to 4500 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12017134 Supplier Cole-Parmer™ Supplier No. 0472901
  • ONLINE EXCLUSIVE PRICE
    356.00€
    307.62€ / pezzi 1
    Risparmia 48.38€
    13% Sconto
View all promotions
Spedizione stimata: 30-12-2026
This V Series Stuart™ Vortex Mixer is a compact unit perfect for individual work stations.
  • Mix a wide variety of tube sizes and shapes with the included rubber cup head.
  • Heavy die-cast body won't dance across your bench.
  • Automatic start when the soft rubber cup is pressed down.
  • The Vortex mixer provides rapid mixing of samples contained in test-tubes, small flasks and bottles.
  • Starts automatically when the rubber cup is depressed and stops once vessel is removed.
  • Control the mixing speed by selecting from 200 rpm for more gentle mixing to 2500 rpm for vigorous agitation.
  • Heavy die-cast body for stability during use.
  • Includes integral retort rod for securing vessels above the vortex action for long-term mixing.
Specifications
Speed Range Fixed Speed
Product Type Vortex Mixer