Descrizione
- Easy to replace, visor snaps into place
- For use with 3M™ V4D Faceshield
- Replacement clear acetate visor offering good chemical resistant properties
- Acetate visor has good optical properties and an anti-fog coating
- Replacement clear polycarbonate visor offering impact and scratch resistance properties
- Replacement steel mesh visor with a black matt finish providing protection from flying chips and splinters
Specifica
Specifica
| Certificazione/Conformità | CE Approved, EN 166 , EN 166 |
| Altezza (sistema metrico) | 0.25 cm |
| Forma | Molding |
| Larghezza (sistema metrico) | 30.48 cm |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | V4 Series Replacement Visor |
| Da utilizzare con (apparecchiatura) | V4F visor |
| Lunghezza (sistema metrico) | 38.1 cm |
| Lens Material | Policarbonato |