Learn More
Greiner Bio-One VACUETTE™ Heparin Blood Collection Tubes
Codice prodotto. 10080348
Cambia vista
Click to view available options
Capacità (sistema metrico):
2 mL
4 mL
6 mL
9 mL
Tipo di chiusura:
Non-ridged Pull Cap
Additivo:
Lithium Heparin
Sodium Heparin
Dimensione della confezione:
pezzi 100
pezzi 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Tipo di chiusura Additivo unitSize
10080348 9 mL Non-ridged Pull Cap Sodium Heparin pezzi 50
13279072 9 mL Non-ridged Pull Cap Sodium Heparin pezzi 100
15737869 2 mL Non-ridged Pull Cap Lithium Heparin pezzi 50
13048762 6 mL Non-ridged Pull Cap Lithium Heparin -
15422560 6 mL Non-ridged Pull Cap Lithium Heparin pezzi 50
15482520 4 mL Non-ridged Pull Cap Lithium Heparin pezzi 50
15206067 9 mL Non-ridged Pull Cap Lithium Heparin pezzi 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Codice prodotto. 10080348 Fornitore Greiner Bio-One N. del fornitore 455051
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Aggiungi al carrello
VACUETTE™ Heparin Blood Collection Tubes have a green top and contain spray-dried lithium, sodium or ammonium heparin on the inner walls and are usedin clinical chemistry, immunology and serology. The anticoagulant heparin activates antithrombin, which blocks the clotting cascade and thus produces a whole blood/plasma sample.
  • Tubes are supplied in packs of 50 pieces
  • For venus blood collection
  • Primarily used for drawing blood for chemistry and immunochemistry testing
  • Made out of virtually unbreakable PET plastic
  • Provides improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • VACUETTE™ Lithium Heparin Blood Collection Tubes are also available with a gel separator
  • Heparin tubes from Greiner Bio-One are available in: LH Lithium Heparin, LH Lithium Heparin Sep and NH Sodium Heparin.
  • CE marked
TRUSTED_SUSTAINABILITY
Specifica
Product Type Blood Collection Tube
Additivo Sodium Heparin
Materiale PET
Sterilità Sterile
Tipo di chiusura Non-ridged Pull Cap
Capacità (sistema metrico) 9 mL
Da utilizzare con (apparecchiatura) VACUETTE™ Blood Collection Needles
Colore chiusura Green with Black Ring
Altezza (sistema metrico) 100 mm
Dimensioni provetta 16 x 100 mm
Da utilizzare con (applicazione) Blood Collection
Diametro (sistema metrico) esterno 16 mm