Learn More
Greiner Bio-One VacuetteTube
Product Code. 15422244
Change view
Click to view available options
Capacity (Metric):
1 mL
2 mL
2.5 mL
4 mL
Closure Color:
Green
Lavender
Unit Size:
Pack of 1200
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
15422244 2 mL Lavender Pack of 50
10667505 4 mL Green Pack of 1200
10607344 1 mL Lavender Pack of 50
10247597 2.5 mL Green Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15422244 Supplier Greiner Bio-One Supplier No. 454087
  • ONLINE EXCLUSIVE PRICE
    10.50€
    9.93€ / pezzi 50
    Risparmia 0.57€
    5% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 26-11-2026
Greiner Bio-One VacuetteTube, Capacity: 4mL, Closure Color: White, Closure Type: Cap, Sterile, Diameter Inside: 13mm, Height: 75mm, Vacuette Tube
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuette Tube
Sterility Sterile
Closure Type Cap
Capacity (Metric) 2 mL
Diameter (Metric) Inner 13 mm
Closure Color Lavender
Height (Metric) 75 mm
compliance-icons
Product Identifier
  • VACUETTE K3E K3EDTA
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediaminetetraacetic Acid Tripotassium Dihydrate