Learn More
Integra Biosciences™ VACUSIP Tubing Set
Product Code. 11787312
Change view
Click to view available options
Includes:
VACUSIP Hand Operator, 1.2m Silicone Tubing, 0.22m Silicone Tubing, 1 Filter (Non-sterile, 0.45μm)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11787312 VACUSIP Hand Operator, 1.2m Silicone Tubing, 0.22m Silicone Tubing, 1 Filter (Non-sterile, 0.45μm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11787312 Supplier Integra Biosciences™ Supplier No. 159040
  • ONLINE EXCLUSIVE PRICE
    51.95€
    45.03€ / pezzi 1
    Risparmia 6.92€
    13% Sconto
View all promotions
Spedizione stimata: 09-12-2026
Specifications
Includes VACUSIP Hand Operator, 1.2m Silicone Tubing, 0.22m Silicone Tubing, 1 Filter (Non-sterile, 0.45μm)
Product Type VACUSIP Tubing Set