Learn More
BD Vacutainer™ Heparin Plasma Tubes
Product Code. 16090902
Change view
Click to view available options
Capacity (Metric):
4 mL
6 mL
10 mL
Additive:
Lithium Heparin
Sodium Heparin
Material:
Glass
PET
Closure Type:
Hemogard
Traditional Cap
Unit Size:
Pack of 100
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Additive Material Closure Type Unit Size
16090902 10 mL Sodium Heparin Glass Traditional Cap Pack of 100
12977676 6 mL Sodium Heparin PET Hemogard Pack of 1000
12957676 6 mL Lithium Heparin PET Hemogard Pack of 1000
12967676 4 mL Sodium Heparin PET Hemogard Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16090902 Supplier BD Supplier No. 368480
  • ONLINE EXCLUSIVE PRICE
    62.50€
    35.56€ / pezzi 100
    Risparmia 26.94€
    43% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 29-12-2026
Achieves optimum anticoagulation by the use of pharmaceutical-grade Heparin. BD Vacutainer™ Heparin Plasma Tubes are made of durable PET plastic with a green Hemogard™ closure. Tubes in 2, 4, 6 and 10mL sizes are available with either Sodium heparin or Lithium heparin as an additive.
BD Vacutainer™ Heparin Plasma Tubes for clinical chemistry are available with spray-dried sodium heparin or lithium heparin additive. Heparin acts as an anticoagulant as it develops an antithrombin complex. This complex inhibits thrombin and the activated factor X and thus prevents coagulation.
  • The optimum anticoagulation is achieved by the use of 17 IU heparin per mL of blood (when the fill level is correct)
  • The heparin is spray dried onto the inner walls of the tubes using a special procedure so that the additive is evenly distributed to achieve the best possible solubility
  • For clinical chemistry, lithium heparin is generally preferred over sodium heparin
  • Correct mixing (8-10 inversions) of the tube immediately after the blood sample has been taken is extremely important to avoid microclotting
  • Centrifugation conditions: ≤1300 g for 10 minutes at 18–25°C
  • The 10mL Sodium Heparin tube is made of glass and features a conventional closure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heparin Plasma Tube
Additive Sodium Heparin
Material Glass
Sterility Sterile
Closure Type Traditional Cap
Capacity (Metric) 10 mL
For Use With (Equipment) Vacutainer™ Needle and Tube Holder
Closure Color Green
Tube Size 16 x 100 mm
For Use With (Application) Used to obtain a plasma sample
Label Type Paper